missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terracycle™ Industrial Safety Equipment Zero Waste Box™

Recycle Industrial Safety Equipment using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.
Brand: Terracycle™ CS199-L-NL
This item is not returnable.
View return policy
Description
Use this Zero Waste Box to recycle:
- Industrial safety helmets
- Safety bump caps
- Hi-vis jackets
- Heavy-duty gloves
- Ear defenders Protective eyewear (goggles)
Do not send in:
- Fabric work gloves with an anti-slip layer
- Suede and leather gloves
- Disposable face masks
- Safety footwear
- Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment, or to people
- Batteries, pressurized canisters, broken glass, and medical waste
Specifications
25 kg | |
Beige, Green, Red, White | |
Single-use | |
Large | |
Not Autoclavable | |
Recycling Box |
55 lb. | |
38 x 38 x 97 cm | |
Yes | |
Non-sterile | |
Cardboard, Plastic |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Terracycle™ Industrial Safety Equipment Zero Waste Box™ > Zero Waste Box; Size: Large
Spot an opportunity for improvement?Share a Content Correction