missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terracycle™ Industrial Safety Equipment Zero Waste Box™

Recycle Industrial Safety Equipment using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.
Marke: Terracycle™ CS199-S-NL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Use this Zero Waste Box to recycle:
- Industrial safety helmets
- Safety bump caps
- Hi-vis jackets
- Heavy-duty gloves
- Ear defenders Protective eyewear (goggles)
Do not send in:
- Fabric work gloves with an anti-slip layer
- Suede and leather gloves
- Disposable face masks
- Safety footwear
- Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment, or to people
- Batteries, pressurized canisters, broken glass, and medical waste
Spezifikation
7 kg | |
Beige, Green, Red, White | |
Single-use | |
Small | |
Not Autoclavable | |
Recycling Box |
15 lb. | |
25 x 25 x 46 cm | |
Yes | |
Non-sterile | |
Cardboard, Plastic |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Terracycle™ Industrial Safety Equipment Zero Waste Box™ > Zero Waste Box; Size: Small
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur