Learn More
Thermo Scientific™ Indiko™ and Konelab™ System D-Dimer to Transferrin Specific Protein Reagents
Choose from a specific protein test menu of over 20 immunoturbidimetric methods with Thermo Scientific™ Indiko™ and Konelab™ System Specific Protein Reagents.
€ 194.00
Description
Receive best-in-class methodologies and standardized performance with Thermo Scientific™ Indiko™ and Konelab™ System Specific Protein Reagents, which include a specific protein test menu of 15 immunoturbidimetric methods. All reagents are ready to go and packaged in volume-optimized, barcoded kits; in-house evaluation as well as optimization of reagents guarantees continuous support and optimum results.
- High-quality reagents are barcoded for automatic identification
- Most reagents are delivered in ready-to-use liquid form
- Special attention has been paid to the optimization of package sizes
- The CE marked products are compliant with EU regulatory requirements for in vitro diagnostic medical devices
Specific Protein Applications for Indiko and Konelab Analyzers:
- Albumin / Microalbuminuria
- Apolipoprotein A1 & B
- Complement C3 & C4
- C-reactive protein
- D-Dimer
- Ferritin
- HbA1c
- IgA, G & M
- Rheumatoid factors
- Soluble transferrin receptor
- Transferrin
Specifications
Serum | |
Reagent |
Safety and Handling
- SpeciCal Protein calibrator, 5x 1ml 35 .
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- EUH208-Contains 2-methyl-2H-isothiazol-3-one hydrochloride. May produce an allergic reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : 2-methyl-2H-isothiazol-3-one hydrochloride
Your input is important to us. Please complete this form to provide feedback related to the content on this product.