Description
- For the secure positioning and leveling of a BINDER™ CO2 incubator
- Suitable for:
- CB 260
- CB-S 260
Specifications
Specifications
| Description | Base - the base equipped with casters is used for the secure positioning and leveling of a BINDER CO2 incubator |
| For Use With (Equipment) | BINDER Chambers Chambers CB260, CB-S260, CBF 260 |
| Material | Steel |
| Type | Base |
| Includes | Casters |
| Dimensions (L x W x H) | 920 x 920 x 350 mm |
| Weight (English) | 18.74 lb. |
| Weight (Metric) | 8.5 kg |