Learn More
Inconel∣r 600 gauze, 200 mesh woven from 0.05mm (0.002in) dia wire, Thermo Scientific Chemicals
€ 57.39 - € 167.84
Specifications
Chemical Name or Material | Inconel™ 600 gauze |
---|---|
Name Note | 200 mesh woven from 0.05mm (0.002 in.) dia. wire |
MDL Number | MFCD01741927 |
Mesh Size | 200 Mesh |
Form | Wire Cloth |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15497305
|
Thermo Scientific Alfa Aesar
046791.FL |
75 x 75 mm |
€ 57.39
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15407315
|
Thermo Scientific Alfa Aesar
046791.GT |
150 x 150 mm |
€ 167.84
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Inconel™ 600 gauze | |
MFCD01741927 | |
Wire Cloth |
200 mesh woven from 0.05mm (0.002 in.) dia. wire | |
200 Mesh |
Safety and Handling
GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350-H372
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only