Learn More
Thermo Scientific™ Impeller Sleeve and Connector for Drive Shaft DS300
Product Code. 30040292 Shop All Thermo Scientific Products
Click to view available options
Capacity (Metric):
50 L
100 L
200 L
For Use With (Equipment):
108 cm (43 in.) Drive Shaft
66 cm (26 in.) Drive Shaft
84 cm (33 in.) Drive Shaft
Unit Size:
Pack of 10
Pack of 12
Pack of 7
Product Code. 30040292
Brand: Thermo Scientific™ SH30749.12
The Impeller sleeve for open-top mixing supports mixing applications in bioproduction using Thermo Scientific™ Bioprocess Containers (BPCs) for Thermo Scientific™ HyPerforma™ Single-Use Mixer, with impeller-based mixing in open or closed designs.
  • For use with the open top BPC tank liner and re-usable hub.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 100 L
For Use With (Equipment) 84 cm (33 in.) Drive Shaft
Type Impeller Sleeve