missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Immuno Clear Standard Modules

Perform applications that deliver well-to-well reproducibility with these immuno clear standard modules. Manufactured to fit standard equipment.

Brand:  Thermo Scientific™ 469264

 View more versions of this product

Product Code. 10578261

This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Breakable Strips
Certified for adsorption of IgG; CV of <5% between wells
Strip Plate
320 Cases
Clear
Polystyrene
250 μL
Round
80
Antibodies (Glycoproteins)
Hydrophilic
Yes
Colorimetric
Adsorption
Microplate Strip
16
Hydrophilic
320
None
U16
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number165232 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number469264
Lot Number177802 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number469264
Lot Number180798 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number469264
Lot Number169845 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number469264
Lot Number171586 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number469264
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Immuno Clear Standard Modules >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.