Learn More
Cytiva Immobiline™ Sample Application Strips
Offer improved gel performance with faster and more stable gradients, fewer artifacts, and easier zymogram assays
Brand:  Cytiva 18-1002-74
264.10 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
Product Code. 10565395
  • € 286.00 / Pack of 5
In Stock
to see stock.
Explore more special offers
Description
  • For use with SDS and immobiline samples
  • Silicone applicator strip with sample holes to load samples onto Isoelectric focussing (IEF) Gels
TRUSTED_SUSTAINABILITY
Specifications
Specifications
26 SDS sample
SDS Sample Application Strip
5