Learn More
Cytiva Ready-To-Go™ GenomiPhi V3 DNA Amplification Kit
Shop All Cytiva Products
Click to view available options
No. of Reactions:
24
96
480
missing translation for 'unitSize'
Each
Beschreibung
p>
- Representative amplification of the whole genome.
- Preformulated, predispensed, single-dose, lyophilized cakes for rapid reaction set-up and greater reproducibility.
- Quick and simple automation-friendly protocol; no thermal cycler required.
- Shipping and storage at room temperature provides easy access and reduced carbon footprint.
- Yields high-quality DNA (e.g. for high-throughput genotyping and DNA archival).
- The reaction mixture is provided as predispensed, single-dose, lyophilized cakes in either strips of 8 tubes, or 96-well plates.
- Amplification is highly uniform over the entire genome so that locus representation remains extremely close to the original DNA sample.
- Amplification is carried out with very high fidelity due to Phi29 DNA polymerase proofreading activity.
- The average product length is over 10kb.
- Procedure is quick, simple and does not require a thermocycler.
- A typical DNA yield of 12 to 20μg DNA can be achieved in less than two hours from only 10ng of genomic DNA input.
- The starting material for GenomiPhi reactions can be purified DNA from any commercial kit or homebrew method, or a nonpurified cell lysate may be used.
- Protocols for the amplification of DNA from blood lysates and dried blood spots are included in the manual.
- GenomiPhi amplified DNA is suitable for various applications such as genotyping (SNP, STR, array CGH), cloning, sequencing, and DNA archival.
Spezifikation
Spezifikation
| Content And Storage | Store the kit at ambient temperature.Once opened, completely reseal the pouch containingthe Ready-To-Go GenomiPhi V3 DNA amplification kit cakes. To ensure maximum shelf life and optimal performance, storage in a desiccator is recommended.Store recons |
| Description | illustra Ready-To-Go GenomiPhi V3 DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 12 to 20 μg yield from nanogram amounts of DNA sample. |
| For Use With (Application) | Genomic DNA Amplification |
| No. of Reactions | 480 |
| Product Type | illustra Ready-To-Go GenomiPhi V3 DNA Amplification Kit 5x96 Reactions |
| Size | > 10 kb |
| Test Time | 1.5-2 h |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur