missing translation for 'onlineSavingsMsg'
Learn More

IGHM Mouse anti-Human, Clone: 2D10B10, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66484-1-IG-20UL

 View more versions of this product

Product Code. 16828405

  • € 145.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.
Specifications

Specifications

IGHM
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P01871
IGHM
IGHM Fusion Protein Ag1459
20 μL
Primary
Human
Antibody
IgG1
Western Blot, Immunohistochemistry (Paraffin)
2D10B10
Unconjugated
IGHM
IGHM, MU, VH
Mouse
Protein G
RUO
3507
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.