Learn More
IGF1 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 500-P11BT-25UG
Description
AA Sequence of recombinant protein: GPETLCGAEL VDALQFVCGD RGFYFNKPTG YGSSSRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPAKSA. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IGF-I. Anti-Human IGF-I-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human IGF-I by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human IGF-I (500-P11) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human IGF-I. Western Blot: To detect hIGF-I by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hIGF-I is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P11BT-1MG will be provided as 2 x 500 μg
IGF1 (Insulin-like growth factor-1) is structurally and functionally related to insulin but has a much higher growth-promoting activity. A variety of cellular responses are induced by IGF1, including cell proliferation, differentiation, migration, and survival. Further, IGF1 is a polypeptide growth factor that stimulates the proliferation of a wide range of cell types in muscle, bone, and cartilage tissue. IGF1 stimulates glucose transport in rat bone-derived osteoblastic (PyMS) cells and is effective at much lower concentrations than insulin, not only regarding glycogen and DNA synthesis but also with regards to enhancing glucose uptake. In circulation, IGFs are predominantly bound to binding proteins (IGFBPs) which prolong the half-life of the IGFs and play a role in delivering them to target tissues. IGF-I is known as one of the most potent activators of the AKT signaling pathway which is known to be a stimulator of proliferation and an inhibitor of programmed cell death. Moreover, mature human IGF-I is 100% homologous with bovine and porcine proteins. Low levels of IGF1 have been linked to Alzheimer's disease. IGF1 is processed from a precursor, bound by a specific receptor, and secreted. Defects in the IGF1 gene are a cause of insulin-like growth factor 1 deficiency and several transcript variants encoding different isoforms have been found.Specifications
IGF1 | |
Polyclonal | |
Biotin | |
IGF1 | |
C730016P09Rik; class 1 insulin-like growth factor I preproprotein; class 2 insulin-like growth factor I preproprotein; H-IGF-1; IBP1; IGF; IGF1; Igf-1; IGF1A; IGFI; Igf-I; IGFIa; IGF-IA; IGF-IB; insulin growth factor-1; Insulin like growth factor; insulin like growth factor 1; insulin-like growth factor 1; insulin-like growth factor 1 (somatomedin C); insulin-like growth factor I; insulin-like growth factor I (somatomedin C); insulin-like growth factor IB; mechano growth factor; MGF; M-IGF-1; OTTHUMP00000195084; prepro-IGF-I; prepro-insulin-like growth factor I; R-IGF-1; somatomedin; Somatomedin C; Somatomedin-C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3479 | |
-20°C |
ELISA, Western Blot | |
0.1-1.0 mg/mL | |
PBS with no preservative | |
P05019 | |
IGF1 | |
E.coli-derived Recombinant Human IGF-I | |
25 μg | |
Primary | |
Human | |
Lyophilized |