Learn More
IBA1 Mouse anti-Human, Unconjugated, Clone: 1C6A10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66827-1-IG-150UL
 View more versions of this product
Product Code. 16819065
  • € 410.00 / 150µL
Description
The IBA1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IBA1. This antibody recognizes human antigen. The IBA1 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
IBA1
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Immunohistochemistry, Immunofluorescence
1C6A10
Unconjugated
150 μL
199
IgG2a