Learn More
Ambion™ mirVana™ miRNA Inhibitor
mirVana™ miRNA Inhibitor
Brand:  Ambion™ 4464088
2623.05 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
 View more versions of this product
Product Code. 13426706
  • € 3915.00 / 250nmol
Explore more special offers
Description
Description
miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store in freezer (-5 to -30°C).
miRNA
miRNA
250 nmol
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Ambion™ mirVana™ miRNA Inhibitor > 250 nmol