Learn More
Thermo Scientific™ HyperSep™ Pressure Manifold Adapter Plates
Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
1 mL Cartridges
6 mL Cartridges
HyperSep Positive Pressure Manifold, 1 mL SPE Cartridges
HyperSep Positive Pressure Manifold, 10 mL/15 mL SPE Cartridges
missing translation for 'unitSize'
Each
Description
Use the Thermo Scientific™ HyperSep™ Positive Pressure Manifold Adapter Plate that matches the size of solid phase extraction (SPE) cartridges used. The HyperSep Positive Pressure Manifold can hold up to 48 SPE cartridges of various sizes: 1, 3, 6, and 10/15mL.
The HyperSep Positive Pressure Manifold holds up to 48 SPE cartridges. Samples and solvent, added to the SPE cartridge, are extracted under positive pressure for an even flow at each position. Extracts are collected into 13 or 16mm tubes. This SPE manifold provides a quick, simple and robust way to prepare multiple samples for analysis by chromatography.
See related products if you need replacements for any of these HperSep Positive Pressure Manifold accessories.
Spécification
Spécification
| Description | Adapter Plate for 10 mL/15 mL Cartridges for PP Manifold, 1 pk |
| For Use With (Application) | Industrial Chromatography |
| For Use With (Equipment) | HyperSep Positive Pressure Manifold, 10 mL/15 mL SPE Cartridges |
| Product Line | HyperSep |
| Type | Adapter Plate |