Learn More
Ansell™ HyFlex™ 11-619 Series Knitted Polyurethane Light Gloves
Product Code. 13488958 Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
144 pairs
Product Code. 13488958
Brand: Ansell™ 11619_8
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Almost a second skin, this very thin glove offers outstanding comfort, fit and sensitivity for the most delicate handling. Ansell HyFlex™ 11-619 Series Knitted Polyurethane Light Gloves feature 18-gauge knitting technology which allows the glove to be ultra-thin yet tough.
  • Designed for a wide range of lightweight applications in dry or slightly oily environments
  • Ideal for workers who need a higher level of fit, touch and precision than that offered by their current gloves
  • Also ideal for applications where cotton gloves or no gloves at all are used despite the risk of scrapes, blisters and minor injuries
  • Ideal for final assembly, airco, electrical engines, wire harness assembly, adjusters work
TRUSTED_SUSTAINABILITY
Specifications
Back Material Polyurethane
Certifications/Compliance CE Mark, EN 388 : 2121
Cuff Material Polyurethane
Coating Material Polyurethane
Product Type Lightweight Gloves
Size 8
Style 11-619
Glove Cuff Style Knit
Lining Nylon
Palm Type Coated-palm
Quantity 144 Pairs
Series 11-619