Learn More
Hydroxy Itraconazole-d8, MedChemExpress
MedChemExpress Hydroxy Itraconazole D8 is the deuterium labeled Hydroxy Itraconazole. Hydroxy Itraconazole is an active metabolite of Itraconazole (ITZ), which is a triazole antifungal agent.
Brand: Medchem Express HY-12772S
This item is not returnable.
View return policy
Description
Hydroxy Itraconazole D8 is the deuterium labeled Hydroxy Itraconazole. Hydroxy Itraconazole is an active metabolite of Itraconazole (ITZ), which is a triazole antifungal agent.Specifications
Hydroxy Itraconazole-d8 | |
White | |
C35H30D8Cl2N8O5 | |
O=C1N(C(C([2H])([2H])[2H])([2H])C(O)([2H])C([2H])([2H])[2H])N=CN1C2=CC=C(N3CCN(C4=CC=C(OCC5OC(CN6N=CN=C6)(C7=CC=C(Cl)C=C7Cl)OC5)C=C4)CC3)C=C2 | |
729.68 | |
Research |
1 mg | |
Research | |
R-63373-d8 | |
729.68 | |
98.0% | |
Powder |
Research purposes only