missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.5M, Chem-Lab™

Product Code. 10365932
Click to view available options
Quantity:
5 L
Packaging:
EP
Unit Size:
5 L
This item is not returnable. View return policy

Product Code. 10365932

Brand: Chem-Lab CL05.0310.5500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For laboratory use, Titrations, ASTM D-94, D-128

Specifications

Appearance Colorless
pH <1
CAS 7647-01-0
Composition Hydrochloric Acid 0.5mol/L
Concentration 18.23g HCl/l H2O = 0.5 N (±0.001/20°C)
Density 1.01g/mL
Odor Odorless
Packaging EP
Normality 0.5N (±0.001 at 20°C)
Quantity 5 L
Molecular Formula ClH
Physical Form Liquid
MDL Number MFCD00011324 MFCD00792839
UN Number UN1789
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid
Solubility Information Soluble in Water
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.46g/mol
Concentration or Composition (by Analyte or Components) 18.23g HCl / 1L H2O
Chemical Name or Material Hydrochloric Acid
Show More Show Less
compliance-icons
Product Identifier
  • Hydrochloric acid 0.5 mol/l
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contains : HCl/H2O
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.