Learn More
Emma Safety Footwear Hydro-Tec™ Stability Inlay Soles
Provides foot with side support and extra support at heel and middle of foot.
Brand: Emma Safety Footwear 1072/41
Description
- Regulates moisture
- Contains active carbon to prevent odor
Specifications
Gray | |
41 | |
Stability Inlay Sole |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.