missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emma Safety Footwear Hydro-Tec™ Stability Inlay Soles
Provides foot with side support and extra support at heel and middle of foot.
Brand: Emma Safety Footwear 1072/39
This item is not returnable.
View return policy
Description
- Regulates moisture
- Contains active carbon to prevent odor
Specifications
Gray | |
39 | |
Stability Inlay Sole |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction