Learn More
Smart Chemical HYDRANAL™ - Titrant 5, Honeywell Fluka™
Titrating agent for volumetric two-component Karl Fischer titration (methanol-based).
Brand: Honeywell Fluka 34801-1L-SC
Description
Honeywell's Smart Chemical Hydranal™ bottles are embedded with an RFID chip, visible in the form of a new label, that captures essential information for every titration, including article code, product name, lot number, shelf life and numeric concentration.
- Secure Data Transfer: Ensure correct and complete reagent data every time with traceable information flow.
- Efficient System Setup: Reduce system set-up time with automated reagent data transfer.
The RFID label is currently compatible with Mettler Toledo's Karl Fischer Compact and Excellence line titrators (when upgraded with external RFID reader) and is available on our best-selling Hydranal Volumetric Titrants.

Specifications
HYDRANAL™-Titrant 5 | |
11°C | |
67-56-1, 7553-56-2 | |
UN1230 |
0.85 g/mL | |
1 L | |
Mettler-Toledo Smart Chemical Bottle |
Safety and Handling
- HYDRANAL NEXTGEN Titrant 5 SC
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : methanol, iodine
Your input is important to us. Please complete this form to provide feedback related to the content on this product.