Learn More
Cytiva HyClone™ Tetracycline Screened Fetal Bovine Serum
Click to view available options
Heat Inactivated:
No
Yes
Unit Size:
500mL
Specifications
Specifications
| Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. |
| Country of Origin | United States |
| Specialty Type | Tetracycline Screened |
| Quantity | 500 mL |
| Sterility | Sterile |
| Sterilization Method | Sequential-filtered, 40 nm |
| Grade | Defined, Specialty screened |
| Tested For | Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus |
| Volume (Metric) | 500 mL |
| Endotoxin Concentration | less than or equal to 10 EU/mL |
| Show More |