Learn More
Cytiva HyClone™ Minimum Essential Medium
Click to view available options
Quantity:
1000 mL
500 mL
6 x 1000 mL
6 x 500 mL
With Additives:
Earle's Balanced Salt Solution (EBSS)
Glutamine
Glutamine, Earle's Balanced Salt Solution (EBSS)
Glutamine, Ribonucleosides, Deoxyribonucleosides
Phenol Red
Unit Size:
1000mL
500mL
Pack of 6
Description
- System tested with Cytiva HyClone sera to ensure product efficacy and homogeneity
- All products are hydrated using purified process water and undergo sterile filtration
- Manufactured using ISO 9001 certified processes
Specifications
Specifications
| Form | Liquid |
| Product Type | MEM with EBSS, L-glutamine |
| Sterility | Sterile-filtered |
| Sterilization Method | 0.1 μm Sterile-filtered |
| With Additives | Glutamine, Earle's Balanced Salt Solution (EBSS) |
| Without Additives | None |
| Quantity | 6 x 500 mL |
| Certifications/Compliance | cGMP, ISO 9001:2000 |
| Content And Storage | 2° to 8°C, away from light |
| Description | MEM with Earle's Balanced Salt Solution (EBSS), with 2.0mM L-Glutamine |