Learn More
Cytiva HyClone™ Hank's Balanced Salt Solution Powder
Click to view available options
Quantity:
1 x 10 L
2 x 5 L
With Additives:
Calcium, Magnesium, Phenol Red
Phenol Red
Unit Size:
10 L
Pack of 2
Description
HyClone™ Hanks' Balanced Salt Solution Powder is available in lot sizes that facilitate scale-up and manufacturing.
Specifications
Specifications
| Certifications/Compliance | cGMP and ISO 9001:2000 Guidelines |
| Content And Storage | 2° to 8° C |
| Description | With Calcium, with Magnesium, with Phenol Red (powder) |
| Form | Powder |
| With Additives | Calcium, Magnesium, Phenol Red |
| Without Additives | None |
| Quantity | 2 x 5 L |