Learn More
Cytiva HyClone™ Hank's Balanced Salt Solution Powder
Product Code. 10474672
Click to view available options
Quantity:
1 x 10 L
2 x 5 L
With Additives:
Calcium, Magnesium, Phenol Red
Phenol Red
Unit Size:
10 L
Pack of 2
Product Code. 10474672
Brand: Cytiva SH30015.03
Available in lot sizes that facilitate scale-up and manufacturing.
HyClone™ Hanks' Balanced Salt Solution Powder is available in lot sizes that facilitate scale-up and manufacturing.
  • Cell culture application
  • All formulations incorporate a rigorous specification and approval process
  • TRUSTED_SUSTAINABILITY
    Specifications
    Certifications/Compliance cGMP and ISO 9001:2000 Guidelines
    Content And Storage 2° to 8° C
    Description With Calcium, with Magnesium, with Phenol Red (powder)
    Form Powder
    With Additives Calcium, Magnesium, Phenol Red
    Without Additives No Sodium Bicarbonate
    Quantity 1 x 10 L