missing translation for 'onlineSavingsMsg'
Learn More

HyClone™ 2 M NaCl, Cytiva

Product Code. 17285906
Click to view available options
Quantity:
1 L
10 L
20 L
50 L
100 L
200 L
Packaging:
Single-use Bioprocessing Container
This item is not returnable. View return policy
This item is not returnable. View return policy

Concentrated sodium chloride solutions for applications in upstream and downstream bioprocessing.

HyClone™ NaCl solutions in single-use bags offer a convenient format for many upstream and downstream bioprocessing applications.

  • Manufactured in ISO 9001 and ISO 13485 certified facility.
  • Lot-specific certificate of analysis (CoA) for each lot produced.
  • Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.
  • 0.2 μm sterile filtered.
  • Provided in single-use bag containers.
  • Raw material is EP, USP, and JP grade.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Color Clear, Colorless
Quantity 1 L
Concentration or Composition (by Analyte or Components) 2 M
Packaging Single-use Bioprocessing Container
Product Line HyClone™
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'