Learn More
Boekel Scientific™ Hybridization Glass Bottles for Boekel Little Shot Oven
GREENER_CHOICE
For use with Boekel Little Shot Oven, these glass hybridization bottles fit the standard carousel and help maintain the perfect temperature and mixing for all applications.
Brand:  Boekel Scientific™ 502-0300
 View more versions of this product
Product Code. 10345393
  • € 200.00 / Pack of 2
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Description
  • Pack of 2 Bottles
  • Comes with Blue Caps
  • For Use with Standard Carousel
Specifications
Specifications
Blue
35 mm
Blue caps and silicone O-ring
300 mm
Hybridization Bottle
1.37 in.
Boekel little SHOT III medium capacity oven
11.8 in.
Glass