Learn More
Rotronic™ Humidity standards
For on-site calibration and adjustment of ROTRONIC probes with handheld instrument
Brand:  Rotronic™ EA10SCS
 View more versions of this product
Product Code. 11960655
Includes: SCS traceable calibration certificate, textile pads, Calibration instructions
Description
Description
With the humidity standards, a calibration device and HW4 software, running on a PC this is easily achieved.
  • Ampoules contain unsaturated salt solutions
  • Simple and safe on-site calibration method
  • Unlimited storage life
Specifications
Specifications
0.1
On-site Calibration and Adjustment of Rotronic Probe
Humidity Standard
SCS traceable calibration certificate, textile pad, calibration instruction
Rotronic Instrument
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Rotronic™ Humidity standards > 10% RH