Learn More
Novus Biologicals™ Human Uterus Fundus Whole Tissue Lysate (Adult Whole Normal)
Brand: Novus Biologicals™ NB820-59277
This item is not returnable.
View return policy
Description
The Human Uterus Fundus Whole Tissue Lysate (Adult Whole Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Western Blot | |
1 mg | |
Total Protein - Human Adult Normal Tissue: Uterus: Fundus | |
Normal | |
Human | |
Human Uterus Fundus Whole Tissue Lysate (Adult Normal) | |
Adult |
Reproductive | |
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors | |
Whole | |
Human | |
5 mg/mL | |
-80°C. Avoid freeze-thaw cycles. | |
Native |
Product Content Correction
Product Title
Novus Biologicals™ Human Uterus Fundus Whole Tissue Lysate (Adult Whole Normal) > 1mg