missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TSC1 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0248
This item is not returnable.
View return policy
Description
TSC1 Synthetic Peptide, PEP-0248, from Invitrogen.
This peptide corresponds to 15 amino acids near the carboxy terminus of human TSC1.PEP-0248 can be used as a blocking peptide with polyclonal antibody PA5-20130.Specifications
7248 | |
purified | |
Human TSC1 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q92574 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
TSC1 | |
0.02% sodium azide | |
50 μg | |
PBS |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction