missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SCF Recombinant Protein
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
For in vitro applications. A dose-response assay should be performed to determine the optimal concentration for specific experiments. Reconstitution and Dilution: Reconstitute lyophilized material in 0.1 mL of 10 mM Acetic acid to a final concentration of 0.1 mg/mL. Dilute in a buffer-containing carrier protein, aliquot, and store at or below -70°C. Avoid multiple freeze-thaw cycles. PBS with 1% BSA or a neutral media containing <10% FCS are suitable buffers.
Stem cell factor (SCF) is the ligand of the c-Kit oncogene and is expressed by various structural and inflammatory cells in the airways. Binding of SCF by the c-Kit receptor leads to homodimerization of the receptor and the activation of signalling pathways such as PI-3, PLC-gamma, Jak/STAT, and MAP kinase pathways. SCF expression leads to the induction of mast cell survival and the expression and release of histamine, pro-inflammatory cytokines and chemokines. The inhibition of the SCF/c-Kit pathway leads to a decrease in histamine levels, mast cell and eosinophil infiltration, IL-4 production and airway hyperresponsiveness, suggesting this pathway may be a useful therapeutic target in inflammatory diseases such as asthma. At least two isoforms of SCF are known to exist.
Specifications
Specifications
| Accession Number | P21583 |
| For Use With (Application) | Bioactivity |
| Formulation | 0.01M acetic acid with no preservative |
| Gene ID (Entrez) | 4254 |
| Molecular Weight (g/mol) | 18.4 kDa |
| Name | Human SCF |
| Purification Method | Purified |
| Quantity | 10 μg |
| Source | E. Coli |
| Storage Requirements | -20°C, Avoid Freeze/Thaw Cycles |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction