Learn More
Novus Biologicals™ Human Serum Albumin Overexpression Lysate
Brand: Novus Biologicals™ NBL1-07449
560.00 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
The Human Serum Albumin Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
The lysate was created in HEK293T cells, using plasmid ID RC25943 and based on accession number NM_477. The protein contains a C-terminal DDK tag. | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. |
Western Blot | |
RIPA buffer | |
Homo sapiens albumin (ALB), mRNA. | |
Native |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 10
Product Content Correction
Product Title
Novus Biologicals™ Human Serum Albumin Overexpression Lysate > 0.1mg