Learn More
Novus Biologicals™ Human Lung Whole Tissue Lysate (Adult Whole Primary Tumor and Normal Paired)
Brand:  Novus Biologicals™ NB820-89077
Product Code. 18287062
  • € 970.00 / 1 set
Explore more special offers
Description
Description
The Human Lung Whole Tissue Lysate (Adult Whole Primary Tumor and Normal Paired) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
Western Blot
1 Set
Matched Pair - Total Protein - Human Primary Tumor and Normal Tissue (PP): Lung
Primary Tumor and Normal Paired
Human
Human Lung Whole Tissue Lysate (Adult Primary Tumor and Normal Paired)
Adult
Respiratory
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Whole
Human
5 mg/mL
-80°C. Avoid freeze-thaw cycles.
Native
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only