missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech™ Human KIM-1/HAVCR1 ELISA Kit
Solid phase sandwich ELISA used to detect and quantify protein levels of endogenous KIM-1
Brand: Proteintech™ KE00136-96T
This item is not returnable.
View return policy
Description
- Recognizes human KIM-1
- KIM-1 (Kidney injury molecule 1, Hepatitis A virus cellular receptor 1, HAVCR, T-cell immunoglobulin, mucin domain 1, TIM-1) is a class I integral membrane glycoprotein with ectodomain containing Ig-like domain and mucin domain
- Acts as membrane receptor for HAV
- Provides costimulatory signal for T cell activation and inhibits development of peripheral tolerance
- Involved in regulation of asthma and allergic diseases
- Reported KIM-1 shed into urine after acute kidney damage
- Marker of renal tubular injury
Process:
- Antibody specific for KIM-1 pre-coated onto microwells
- KIM-1 protein in samples captured by the coated antibody after incubation
- After extensive washing, another antibody specific for KIM-1 is added to detect captured KIM-1 protein
- For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent
- Solution containing sulfuric acid used to stop color development and color intensity
- Proportional to quantity of bound protein, measurable at 450 nm with correction wavelength set at 630 nm
Specifications
BC013325 | |
1.1 pg/mL | |
Sandwich ELISA, Double Antibody | |
Plasma, Serum | |
Human | |
Human | |
<10% | |
Blue ice | |
4.5 hr. |
31.25 to 2000 pg/mL | |
ISO 13485 | |
Coated ELISA Kit | |
DAB | |
26762 | |
Solid | |
<10% | |
-20°C to +4°C | |
Q96D42 |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 10
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 96TEST Human N-cadherin ELISA Kit
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P234-Keep only in original container.
- P264-Wash skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P330-Rinse mouth.
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Proteintech™ Human KIM-1/HAVCR1 ELISA Kit > Human KIM-1/HAVCR1 ELISA Kit
Spot an opportunity for improvement?Share a Content Correction