Learn More
Invitrogen™ Human KCTD15 Synthetic Peptide
Product Code. 15329050
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Product Code. 15329050
Brand: Invitrogen™ PEP0817
Synthetic Peptide
KCTD15 Synthetic Peptide, PEP-0817, from Invitrogen.
Childhood and adult obesity in the United States and to a lesser extent the rest of the world has increased dramatically over the past decade. Both environmental and genetic factors are involved in the onset and progression of weight gain. Recently, the potassium channel KCTD15 was identified as a genetic loci associated with higher than normal body mass index (BMI) in humans along with genes such as GNPDA2, MTCH2, FTO, and TMEM18. Further studies on single nucleotide polymorphisms (SNPs) in non-diabetic and diabetic patients showed that FTO was most strongly associated with obesity while MTCH2 and GNPDA2 were still significantly associated with higher than normal BMI levels. At least two isoforms of KCTD15 are known to exist.
TRUSTED_SUSTAINABILITY
Specifications
Gene ID (Entrez) 79047
Species Human
Purification Method purified
Accession Number Q96SI1
Common Name Human KCTD15
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol KCTD15
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS