missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human HOXD1 (aa 129-207) Control Fragment Recombinant Protein

Product Code. 30199465
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30199465

Brand: Invitrogen™ RP105976

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (84%), Rat (84%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65257 (PA5-65257. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. HOXA1 is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. The encoded protein may be involved in the placement of hindbrain segments in the proper location along the anterior-posterior axis during development. Two transcript variants encoding two different isoforms have been found for this gene, with only one of the isoforms containing the homeodomain region.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9GZZ0
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3231
Name Human HOXD1 (aa 129-207) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias BSAS; homeo box 4 G; homeo box A1; homeo box D1; homeobox 1 F; homeobox A1; homeobox D1; homeobox protein Hox-4.9; Homeobox protein Hox-D1; homeobox protein Hox-GG; HO x 1; HO x 1 F; HO x 2 I; HO x 4; Hox-4.7; Hox-4.9; HO x 4 G; HOXA1; Hoxd1; Hoxd-1; MGC45232
Common Name HOXD1
Gene Symbol HOXD1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GSHVHYATSAVFSGGGSFLLSGQVDYAAFGEPGPFPACLKASADGHPGAFQTASPAPGTYPKSVSPASGLPAAFSTFEW
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.