Learn More
Gibco™ Human Fibronectin Native Protein
Human Fibronectin Native Protein
Brand:  Gibco™ PHE0023
Product Code. 10486873
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Human Fibronectin Native Protein for Western Blot, IHC, ELISA, Ctrl, FN
Natural human fibronectin is intended for use in cell culture applications. Fibronectin is involved in a number of cellular processes including cell adhesion, migration, morphology, and regulation of cell growth and differentiation.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
Shipping Condition: Dry Ice.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cell Culture
Human
Dry Ice
Extracellular Matrix & Cell Adhesion, Stem Cell Research
Fibronectin Natural Human
Store in freezer (-5 to -30°C).
Fibronectin
Gibco™
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only. Not for use in diagnostic procedures.