Learn More
Novus Biologicals™ Human Eye Whole Tissue Lysate (Adult Whole Diabetes)
Brand: Novus Biologicals™ NB820-59288
This item is not returnable.
View return policy
Description
The Human Eye Whole Tissue Lysate (Adult Whole Diabetes) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.Specifications
Immunoprecipitation, Western Blot | |
1 mg | |
Total Protein - Human Diabetic Diseased Tissue: Whole Eye | |
Diabetes | |
Human | |
Human Eye Whole Tissue Lysate (Adult Diabetes) | |
Adult |
Visual | |
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors | |
Whole | |
Human | |
5 mg/mL | |
-80°C. Avoid freeze-thaw cycles. | |
Native |
For Research Use Only