missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ENaC alpha (aa 20-42) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP088
This item is not returnable.
View return policy
Description
Human alpha-ENaC (residues 20-42) Synthetic Peptide, PEP-088, from Invitrogen.
PEP-088 is a 24 amino acid synthetic peptide whose sequence corresponds to residues 20-42 of human alpha-ENaC with a carboxy-terminal cysteine added for carrier protein conjugation. The sequence of this peptide is (amino to carboxy terminus): L (20)-M-K-G-N-K-R-E-E-Q-G-L-G-P-E-P-A-A-Q-Q-P-T (42)-C.This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and endogenous alpha-ENaC, catalog # PA1-920A. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
6337 | |
purified | |
Human ENaC alpha (aa 20-42) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
P37088 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
SCNN1A | |
No preservative | |
50 μg | |
L(20) -M-K-G-N-K-R-E-E-Q-G-L-G-P-E-P-A-A-Q-Q-P-T(42)-C |