missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CTTNBP2 (aa 951-1098) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60789 (PA5-60789. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
CTTB2 is a protein with six ankyrin repeats and several proline-rich regions. A similar gene in rat interacts with a central regulator of the actin cytoskeleton.
Specifications
Specifications
| Accession Number | Q8WZ74 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 83992 |
| Name | Human CTTNBP2 (aa 951-1098) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 3010022N24Rik; 4732477G22Rik; 6430526E05; 9130022E09Rik; AU040881; brain specific cortactin-binding protein CBP90; C7orf8; Cbp90; cortactin binding protein 2; cortactin-binding protein 2; cortactin-binding protein 2 intron retention form; CORTBP2; Cttnbp2; CTTNBP2 intron retention form; KIAA1758; mKIAA1758; ORF4 |
| Common Name | CTTNBP2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction