Learn More
R&D Systems™ Human CD1a (AAA51931) VersaClone cDNA
The CD1 family members are thought to differ in their cellular localization and specificity for particular lipid ligands
Brand: R&D Systems™ RDC1192
This item is not returnable.
View return policy
Description
The Human CD1a (AAA51931) VersaClone cDNA clone contains the complete ORF for CD1a, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CD1a insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
AAA51931 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
10 μg at 0.2 μg/μL | |
997bp | |
Human |
Product Content Correction
Product Title
For Reasearch Use Only; Not for Use in Humans