Learn More
Novus Biologicals™ Human Brain Occipital Lobe Whole Tissue Lysate (Adult Whole Parkinson's)
Brand: Novus Biologicals™ NB820-59413
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The Human Brain Occipital Lobe Whole Tissue Lysate (Adult Whole Parkinson's) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.Spécification
Western Blot | |
1 mg | |
Total Protein - Parkinsons Disease: Brain: Occipital Lobe | |
Parkinson's | |
Human | |
Human Brain Occipital Lobe Whole Tissue Lysate (Adult Parkinson's) | |
Adult |
Nervous | |
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors | |
Whole | |
Human | |
5 mg/mL | |
-80°C. Avoid freeze-thaw cycles. | |
Native |
For Research Use Only