missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech™ Human BMP7 ELISA Kit
Shop All Proteintech Products
Click to view available options
Target Species:
Human
Unit Size:
96 tests
Description
- Recognizes human BMP7
- BMP7 (osteogenic protein-1, OP-1) plays key role in transformation of mesenchymal cells into bone and cartilage
- Glycosylated disulfide-linked 36 kDa homodimeric protein
- Role in induction, development and regulation of adipocytes (BAT)
- Expressed in brain, kidneys and bladder
- Present in cancers (breast, prostate and colon) implicated in regulating cancer cell proliferation
- Overexpression of BMP7 mRNA in colorectal cancer patients significantly associated with poor prognosis and low survival
- High expression level is biomarker for poor prognosis for HCC
Process:
- Antibody specific for BMP7 pre-coated onto microwells
- BMP7 protein in samples captured by the coated antibody after incubation
- After extensive washing, another antibody of biotinylated specific for human BMP7 is added to detect captured human BMP7 protein
- For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent
- Solution containing sulfuric acid used to stop color development and color intensity
- Proportional to quantity of bound protein, measurable at 450 nm with correction wavelength set at 630 nm
Specifications
Specifications
| Accession Number | BC008584 |
| Assay Range | 15.6 to 1000 pg/mL |
| Assay Sensitivity | 0.8 pg/mL |
| Certifications/Compliance | ISO 13485 |
| Detection Method | Sandwich ELISA, Double Antibody |
| Immunoassay Kit Format | Coated ELISA Kit |
| Sample Type | Plasma, Serum |
| Substrate | DAB |
| Target Species | Human |
| Gene ID (Entrez) | 655 |
| Show More |
Safety and Handling
Product Identifier
- 96TEST Human BMP7 ELISA Kit
- Warning
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P234-Keep only in original container.
- P264-Wash skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P330-Rinse mouth.
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction