Learn More
R&D Systems™ Human AMIGO3 (NP_942015) VersaClone cDNA
Amphoterin-induced Gene and ORF 3
Brand:  R&D Systems™ RDC1793
Product Code. 18706608
  • € 747.00 / 10µg
Explore more special offers
Description
Description
The Human AMIGO3 (NP_942015) VersaClone cDNA clone contains the complete ORF for AMIGO3, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The AMIGO3 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
Specifications
Specifications
cDNA
NP_942015
cDNA is provided in 10mM Tris-Cl, pH 8.5
386724
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
AMIGO3
10 μg at 0.2 μg/μL
1528bp
adhesion molecule with Ig-like domain 3, ALI3, Alivin-3, AMIGO-3, amphoterin-induced gene and ORF 3, amphoterin-induced protein 3, KIAA1851, MGC120552
Human
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers