missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human AFP Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP1118
This item is not returnable.
View return policy
Description
AFP Synthetic Peptide, PEP-1118, from Invitrogen.
AFP (Alpha Fetoprotein) is a major plasma protein produced by the yolk sac, intestinal tract and the liver during fetal life. AFP expression in adults is often associated with hepatoma or teratoma. However, hereditary persistence of AFP may also be found in individuals with no obvious pathology. The protein is thought to be the fetal counterpart of serum albumin, and AFP and albumin genes are present in tandem in the same transcriptional orientation on chromosome 4. AFP is found in monomeric, dimeric, and trimeric forms. AFP can also bind to copper ions, nickel ions, fatty acids and bilirubin. The level of AFP in amniotic fluid is used to measure renal loss of protein to screen for spina bifida and anencephaly. High level of serum AFP has been identified in patients with hepatocellular carcinomas (HCC), teratoblastoma, colorectal cancer, pancreatic cancer, and germ cell neoplasms. This peptide corresponds to 14 amino acids near the center of human AFP.PEP-1118 can be used as a blocking peptide with polyclonal antibody PA5-21004.Specifications
174 | |
purified | |
Human AFP | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
P02771 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
AFP | |
0.02% sodium azide | |
50 μg | |
PBS |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction