Learn More
Applied Biosystems™ HT ExoSAP-IT™ High-Throughput PCR Product Cleanup
HT ExoSAP-IT High-Throughput PCR Product Clean-up is an alternative formulation of ExoSAP-IT reagent specifically designed for the unique requirements of high-throughput, automated platforms, and multi-channel pipettes.
Brand: Applied Biosystems™ 783955000RX
2703.46 EUR valid until 2024-12-14
Use promo code "21632" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21632” to get your promotional price
Description
- One-tube PCR clean-up — Add HT ExoSAP-IT reagent directly to PCR product
- Exceptional accuracy — Achieve high quality data even with long read lengths
- 100% sample recovery — No loss of PCR products regardless of the fragment size
- Simple, single-step — Replaces the multiple steps and wasted time inherent with beads or columns
- High-throughput processing — Even faster time to results with a low viscosity formulation, allowing robotic pipetting
- Scalable - Treat reaction volumes from 5 μL to 5 L
- Convenient packaging — Available in 8-tube strips and 12 strips in a 96-well plate
- Highly stable — Retains full functional activity at 4°C for one week and at 25°C for 8 hours
Specifications
Shipped on dry ice. Store at -20°C. | |
PCR Purification | |
PCR Product Clean-up |
PCR Product Clean-up, 5000 Reactions, Single Step, High-throughput Processing, Scalable, Specifically Designed for the Unique Requirements of High-throughput, Automated Platforms and Multi-channel Pipettes | |
5000 Reactions | |
10 mL |
For Research Use Only. Not for use in diagnostic procedures.