missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ HS 024
GREENER_CHOICE

Product Code. 18718726
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18718726

Brand: Tocris Bioscience™ 1832/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly potent MC4 receptor antagonist

HS 024 is a highly potent melanocortin MC4 receptor antagonist (Ki values are 0.29, 18.6, 5.45 and 3.29 nM for cloned human MC4, MC1, MC3 and MC5 receptors respectively). Increases food intake, and blocks α-MSH- and MTII-induced hypotension and bradycardia in rats, following central administration in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Purity 0.95
Content And Storage Store at -20°C
Quantity 1 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.