Learn More
Thermo Scientific™ hPSC Scorecard Assay Service
Shop All Thermo Scientific Products
Click to view available options
:
1 Sample
Description
- A fast and cost-effective alternative to teratoma formation for verification of trilineage differentiation potential in human pluripotent stem cell lines
- This service leverages the TaqMan hPSC Scorecard Panel (A15872), consisting of 94 predefined TaqMan Gene Expression assays (including endogenous controls) dried-down in the wells
- The panel consists of genes associated with the following developmental states: pluripotency, ectoderm, endoderm, and mesoderm
The hPSC Scorecard Assay Service offers:
- Fast data delivery – turnaround time of four weeks or less
- Convenient sample prep – just send a frozen cell pellet
- Quantitative results – compare scores to reference set of well-characterized pluripotent stem cell lines
- Peer endorsement – cited >240 times in scientific literature
- Exceptional support – a dedicated project manager for frequent project communication
Specifications
Specifications
| Quantity | 1 sample |
| Duration | 3 Weeks |
| Product Type | hPSC Scorecard Assay Service |
| Product Line | Applied Biosystems, QuantStudio |
| Field | Research Use Only |
| Target Audience | Stem Cell Researchers |
| Species | Human |