Learn More
SCAT™ HPLC Waste Sets, V2.0
Shop All SCAT Europe Products
Click to view available options
Capacity (Metric):
2.50 L
5 L
10 L
12 L
20 L
30 L
Material:
HDPE
HDPE-EC
Polypropylene
UN-X HDPE
Thread Size:
GL45
S50
S51
S55
S60/61
S70/71
S90
No. of Ports:
3 (2 Capillary, 1 Tubing)
5 (4 Capillary, 1 Tubing)
6 (3 Capillary, 3 Tubing)
Unit Size:
1 set
Each
Description
SCAT™ offers a complete system for safe supply and disposal of solvents in analytics. All products are compatible across generations and perfectly matched to each other.
- Customized system with high-quality set components
- Safety canisters made of HDPE or polypropylene material
- Attractive price compared to single purchase
- Ready to use, no further components necessary
Specifications
Specifications
| For Use With (Application) | HPLC Systems |
| Type | Complete Solvent Collection System |
| Solvent Channels | 3 |
| Material | HDPE |
| Includes | Safety Waste Cap, 2 x PFA fitting 2.3 mm OD, 2 x PFA fitting 3.2 mm OD, 1 x PP tube connector 5 to 11.5 mm ID, 1 x GL14 connection for exhaust filter, 1 x canister, S55, 5 L, PE-HD, 1 x exhaust filter S, with splash guard and change indicator |
| Color | White |
| Thread Size | S55 |
| Capacity (Metric) | 5 L |
| No. of Ports | 3 (2 Capillary, 1 Tubing) |