Learn More
Epredia™ HP35n Ultra High-profile Disposable Microtome Blades
Shop All Epredia Products
Click to view available options
Product Type:
High Profile Blade
Unit Size:
Pack of 50
Description
- Ultra High-profile
- Non-coated
- Highest Durability
Specifications
Specifications
| Product Type | High Profile Blade |
| Profile | High |
| For Use With (Equipment) | Cryostat |
| Disposable | Single-use |
| Coating | None |
| Cutting Angle | 35° |