Learn More
Horst™ Hotbands
Click to view available options
Voltage:
77 V
120 V
230 V
Wattage:
26w
62w
66w
70w
80w
Unit Size:
Each
Specifications
Specifications
| Dimensions (L x W) | 1.13m |
| Dimensions (L x W x H) | 1.13m |
| Voltage | 230 V |
| Electrical Requirements | 230V Voltage |
| Wattage | 62w |
| Product Type | Hotband |